man
Test
chevron-down
Select Language
English
German
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Mixers
Mortars and Pestles
Giese™ Agate Pestle
11786794
Print
Giese™ Agate Pestle
Length: 75mm
Brand:
Giese™ PESTLE75
View more versions of this product
Product Code.
11786794
45.00 €
/ Each
Quantity
Estimated Shipment: 24-10-2024
Log in
to see stock.
Explore more special offers
Add to basket
This item is not returnable.
View return policy
Description
Specifications
Product Suggestions
Videos
SDS
Documents
Certificates
Promotions
Specifications
Length (Metric)
75 mm