Learn More
3M™ Air Supply Tube C253
Product Code. 15759827
Click to view available options
:
Color: Black
Product Code. 15759827
Brand: 3M™ 7000088344
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
3M™ Air Supply Tube C253 for use with Versaflo™ Supplied Air System S-200+
  • Compressed air supply hose with coupling for use with Versaflo™ Supplied Air System S-200+
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE Approved, EN13463-1 IIB 2GD, EN14594:2005 Class 3A/B / Class 4A/B
Connection Type Belt-Mounted
Color Black
Pressure 3.5 to 7.0 bar, 4.5 to 7.0 bar (with Auxiliary air tool connected)
Length (Metric) 30 m