Learn More
Emma Safety Footwear Alaska XD Safety Shoes
Offers exceptional comfort while the steel toe cap protects from falling objects. Easy twist sole provides a 'sticky grip'.
133.00 €
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15818787
|
Emma Safety Footwear
MM30356637 |
37 |
133.00 €
1 pair |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
15828787
|
Emma Safety Footwear
MM30356638 |
38 |
133.00 €
1 pair |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
15838787
|
Emma Safety Footwear
MM30356639 |
39 |
133.00 €
1 pair |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
15848787
|
Emma Safety Footwear
MM30356640 |
40 |
133.00 €
1 pair |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
15858787
|
Emma Safety Footwear
MM30356641 |
41 |
133.00 €
1 pair |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
15868787
|
Emma Safety Footwear
MM30356642 |
42 |
133.00 €
1 pair |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
15878787
|
Emma Safety Footwear
MM30356643 |
43 |
133.00 €
1 pair |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
15888787
|
Emma Safety Footwear
MM30356644 |
44 |
133.00 €
1 pair |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
15898787
|
Emma Safety Footwear
MM30356645 |
45 |
133.00 €
1 pair |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
15808797
|
Emma Safety Footwear
MM30356646 |
46 |
133.00 €
1 pair |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
15818797
|
Emma Safety Footwear
MM30356647 |
47 |
133.00 €
1 pair |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
15828797
|
Emma Safety Footwear
MM30356648 |
48 |
133.00 €
1 pair |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
15838797
|
Emma Safety Footwear
MM30356649 |
49 |
133.00 €
1 pair |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
Description
- S3-Safety shoe with sole plate for wet work
- Oil Nubuck construction
- GripForce™ Easy Twist Outsole
- 3M™ Reflective Striping
- Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
Alaska XD | |
Oiled Nubuck |