Learn More
Alpha-Estradiol, MedChemExpress
MedChemExpress Alpha-Estradiol is a weak estrogen and a 5α-reductase inhibitor which is used as a topical medication in the treatment of androgenic alopecia.
Brand: Medchem Express HY-B0141A

Description
Alpha-Estradiol is a weak estrogen and a 5α-reductase inhibitor which is used as a topical medication in the treatment of androgenic alopecia.Specifications
Alpha-Estradiol | |
White | |
C18H24O2 | |
Alfatradiol Epiestradiol Epiestrol | |
C[C@@]12[C@H](O)CC[C@@]1([H])[C@]3([H])CCC4=C(C=CC(O)=C4)[C@@]3([H])CC2 | |
272.38 | |
Research | |
COVID-19-immunoregulation |
57-91-0 | |
Research | |
500 mg | |
DMSO : 62.5 mg/mL (229.46 mM; ultrasonic and warming and heat to 60°C) | |
272.38 | |
99.52% | |
Powder |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only