missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methylacyl-CoA Racemase/AMACR Antibody (AMACR/8350R), FITC, Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-20847F

Product Code. 18631764

  • 729.00 € / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

alpha-Methylacyl-CoA Racemase/AMACR Monoclonal antibody specifically detects alpha-Methylacyl-CoA Racemase/AMACR in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

alpha-Methylacyl-CoA Racemase/AMACR
Monoclonal
FITC
2-methylacyl-CoA racemase, alpha-methylacyl-CoA racemase, CBAS4, EC 5.1.99.4, RACE, RM
Recombinant full-length human alpha-Methylacyl-CoA Racemase/AMACR protein
0.1 mL
Lipid and Metabolism, Prostate Cancer, Signal Transduction
23600
Store at 4°C in the dark.
IgG κ
Immunohistochemistry (Paraffin)
AMACR/8350R
PBS
Rabbit
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
alpha-Methylacyl-CoA Racemase/AMACR Antibody (AMACR/8350R), FITC, Novus Biologicals™ > 0.1 mL, FITC

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.