Learn More
Ansell AlphaTec™ 38-628 Butyl/Viton Immersion Gloves
AlphaTec™ Gloves 38-628 offer a combination of butyl and viton layer of 0.7 mm for a maximum protection against hazardous chemicals. Provide the best resistance to the most aggressive chemicals without compromising dexterity or comfort. Offer superior protection against aliphatic, halogenated and aromatic hydrocarbons (benzene, toluene, xylene), as well as concentrated mineral acids.
Brand: Ansell 38-628_10
View more versions of this product
This item is not returnable.
View return policy
Description
- Surprisingly flexible and provides excellent dexterity
- Designed for comfort with a natural, curved ergonomic shape and feel
- Offers easy donning and a good grip
- Absence of latex means there's no risk of latex allergies
- Ideal for the chemical, automotive and printing industries, oil and petrol refining, mining, aerospace and disaster response
Specifications
Viton/Butyl | |
Viton/Butyl Polymer | |
Yes | |
10 | |
28 mil | |
Beaded/Rolled | |
Yes | |
1 Pair | |
38-628 |
Viton/Butyl | |
Yes | |
Immersion Gloves | |
38-628 | |
Black | |
Black | |
Individually wrapped bags with insert | |
Highly chemical resistance |