Learn More
Bochem™ Aluminum Dressing Jar
Brand: Bochem™ 8760
This item is not returnable.
View return policy
Specifications
Aluminum | |
Dressing Jar | |
60 mm |
Round | |
20 mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Katalogseite
File size: 613.2 kB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bochem™ Aluminum Dressing Jar > Dia.: 60mm; Height: 20mm
Spot an opportunity for improvement?Share a Content Correction