Learn More
Wesbart™ Aluminum Horizontal Freezer Rack
Product Code. 16224941
Click to view available options
Holds:
Boxes (10/Rack)
Boxes (12/Rack)
Boxes (25/Rack)
Boxes (28/Rack)
Boxes (4/Rack)
Loose Items or Cryoboxes
Positions:
2 Trays/Rack
3 Trays/Rack
4 Trays/Rack
4 Trays/Rack (Not Included)
5 Trays/Rack
7 Trays/Rack
Dimensions (L x W x H):
560 x 140 x 235 mm
560 x 140 x 415 mm
560 x 140 x 420 mm
560 x 145 x 235 mm
640 x 165 x 240 mm
675 x 155 x 290 mm
685 x 140 x 287 mm
688 x 140 x 300 mm
Unit Size:
Each
Product Code. 16224941
Brand: Wesbart™ BH115/2NMT
Please to purchase this item. Need a web account? Register with us today!
Wesbart™ Aluminum Horizontal Freezer Racks are front loading with trays for quick access to cryoboxes and other stored items.
  • Aluminum front access storage rack for tray units (ordered separately, Cat. No. 17311849) for upright freezers
  • For standard cryobox or other freezer storage
  • Only one tray has to be removed from the rack at a time for sample access, minimizing temperature impact on remaining items
  • Range of sizes and capacities available
Specifications
Product Type Freezer Storage Rack
Holds Boxes (10/Rack)
Material Aluminum
Positions 2 Trays/Rack
For Use With (Equipment) Upright Freezers
Dimensions (L x W x H) 685 x 140 x 287 mm