Learn More
Cole-Parmer™ Aluminum Tensioning Tools
Adjust tension from light to very tight using a knob at the base of this tool. Cole-Parmer™ Aluminum Tensioning Tool cuts ties automatically when preset tension is reached. Tension knob features lock to ensure repeatable tension, tie after tie.
Brand: Cole-Parmer™ 06830-00
389.30 EUR valid until 2024-09-30
Use promo code "21675" to get your promotional price.
This item is not returnable.
View return policy
Description
Adjust tension from light to very tight using a knob at the base of this tool. Cole-Parmer™ Aluminum Tensioning Tool cuts ties automatically when preset tension is reached. Tension knob features lock to ensure repeatable tension, tie after tie.
Shock-resistant tool absorbs all impact. Ideal for 18 to 175 lb. cable or zip ties. No twisting necessary.Specifications
Tensioning Tools | |
Aluminum | |
1 Ea. |
Cable, Zip Ties, Aluminum Gun, Stiffer | |
The Aluminum Gun, Stiffer Is Recommended For Jobs in Large Series | |
−40° to +85°C |