missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente™ Amber Erlenmeyer Flasks with Ground Joint

DURAN™ glass Erlenmeyer flask with conical ground joint

Brand:  Lenz Laborglasintrumente™ 03023052

Product Code. 11903381

  • 782.00 € / Pack of 10
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Acc. to DIN EN ISO 4797 and in addition to standard
  • With scale (approx. volume / mL)
  • With inscription label
  • Material: DURAN™
Specifications

Specifications

300 mL
10
87 mm
Borosilicate Glass (3.3)
29/32
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lenz Laborglasintrumente™ Amber Erlenmeyer Flasks with Ground Joint > Capacity (Metric): 300 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.