Learn More
BRAND™ Amber USP Measuring Flask
Product Code. 15831208 Shop All Brand Products
Click to view available options
Capacity (Metric):
5 mL
20 mL
250 mL
1000 mL
Unit Size:
Each
Product Code. 15831208
Brand: BRAND™ 957483
Only null left
Add to basket
Add to basket
With polypropylene stopper
  • Boro 3.3
  • To DIN EN ISO 1042
  • Calibrated to contain (TC, In)
  • With USP batch certificate
  • TRUSTED_SUSTAINABILITY
    Specifications
    Closure Material Polypropylene
    Color Amber
    Capacity (Metric) 20 mL
    Material Borosilicate Glass (3.3)
    Socket Joint Ground Joint NS 10/19
    Class A