missing translation for 'onlineSavingsMsg'
Learn More

Ammonium bromide, Puratronic™, 99.999% (metals basis)

Product Code. p-7049127
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11322078

Brand: Thermo Scientific Alfa Aesar 012981.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12124-97-9 | BrH4N | 97.94 g/mol

Ammonium bromide is used in pharmaceuticals for its bromide ion. It is used for a variety of analytical chemistry applications. It is also used for photography in films, plates and papers; in fireproofing of wood; in lithography and process engraving and as corrosion inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12124-97-9
Molecular Formula BrH4N
Molecular Weight (g/mol) 97.94
MDL Number MFCD00011419
InChI Key SWLVFNYSXGMGBS-UHFFFAOYSA-N
IUPAC Name ammonium bromide
SMILES [NH4+].[Br-]

Specifications

Melting Point 452°C (sublimation)
Color White
Physical Form Crystalline
Quantity 5 g
Merck Index 14,505
Solubility Information Soluble in water (970mg/ml at 25°C),methanol,ethanol,alcohol (freely),and acetone (freely).
Formula Weight 97.97
Percent Purity 99.999%
Grade Puratronic
Sensitivity Hygroscopic
Density 2.429 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium bromide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.