Learn More
Ammonium Chloride Buffer, Reagecon™ Product Code.: 11454779
compliance-icons compliance-icons
Ammonium Chloride Buffer, Reagecon™
BIOPHARMA
Product Code. 11454779 Shop All Reagecon Products
1 L
Click to view available options
Quantity:
1 L
Product Code. 11454779
Brand: Reagecon 4007300
Please to purchase this item. Need a web account? Register with us today!
Specifications
pH 10
Quantity 1 L
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number473025C1 Certificate TypeCertificate of Analysis Date08/04/2025 Catalog Number4007300
Lot Number473023J1 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog Number4007300
Lot Number473022L1 Certificate TypeCertificate of Analysis Date14/06/2023 Catalog Number4007300
Lot Number473021C1 Certificate TypeCertificate of Analysis Date25/05/2022 Catalog Number4007300
compliance-icons compliance-icons
Product Identifier
  • AMMONIUM CHLORIDE BUFFER SOLUTION pH 10.00
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : ammonia, ammonium chloride
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ammonium Chloride Buffer, Reagecon™ >