Learn More
STEMCELL Technologies Ammonium Chloride Solution
Reagent for lysis of red blood cells
82.35 € - 236.25 €
Specifications
For Use With (Application) Lysis of Red Blood Cells
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17159311
Encompass_Preferred
View Documents
STEMCELL Technologies
07800
100 mL
82.35 €
100mL
View alternative products
Save Up to 
»
Estimated Shipment: 29-11-2024
to see stock.
Add to basket
15570226
Encompass_Preferred
View Documents
STEMCELL Technologies
07850
500 mL
236.25 €
500mL
View alternative products
Save Up to 
»
Estimated Shipment: 29-11-2024
to see stock.
Add to basket
Description
Description
  • Ammonium Chloride Solution is recommended for the lysis of red blood cells (RBCs) in preparations of human and mouse peripheral blood, spleen, or bone marrow cells
  • It is buffered and optimized for gentle lysis of erythrocytes, with minimal effect on leukocytes
  • It does not contain a fixative agent, therefore leukocytes are viable following RBC lysis
Specifications
Specifications
Lysis of Red Blood Cells