missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPSO, 0.2M buffer soln., pH 9.5, Thermo Scientific™
35.40 € - 63.90 €
Specifications
CAS | 102029-60-7 |
---|---|
Color | Colorless |
Concentration | 0.2 M |
pH | 9.5 |
Physical Form | Liquid |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
102029-60-7 | |
0.2 M | |
Liquid | |
249.257 |
Colorless | |
9.5 | |
Odorless |
Safety and Handling
Recommended Storage : Keep cold
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
AMPSO, 0.2M buffer soln., pH 9.5, Thermo Scientific™