Learn More
ANA-12, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
ANA-12 is a potent and selective TrkB antagonist with IC50s of 45.6 nM and 41.1 μM for the high and low affinity sites, respectively.
Specifications
Specifications
| Chemical Name or Material | ANA-12 |
| CAS | 219766-25-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H21N3O3S |
| Quantity | 50 mg |
| Solubility Information | DMSO : 9.09 mg/mL (22.31 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (ultrasonic) (insoluble) |
| SMILES | O=C(C1=CC2=CC=CC=C2S1)NC3=CC=CC=C3C(NC4C(NCCCC4)=O)=O |
| Molecular Weight (g/mol) | 407.49 |
| Formula Weight | 407.49 |
| Show More |