Learn More
ANAPC2 Monoclonal Antibody (OTI1H8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503407
Description
APC2 is a cullin-related subunit of the anaphase-promoting complex (APC), or cyclosome. APC2 specifically interacts with APC11, another APC subunit.Specifications
| ANAPC2 | |
| Monoclonal | |
| 0.38 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8BZQ7, Q9UJX6 | |
| Anapc2 | |
| Full length human recombit protein of human ANAPC2 produced in HEk293 cell. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1H8 | |
| Unconjugated | |
| ANAPC2 | |
| APC2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 296558, 29882, 99152 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.