missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ANKRA2 HEK293 Cell Transient Overexpression Lysate (Nondenatured)
Human overexpression lysate
Brand: Abnova™ L014T6.100ug
This item is not returnable.
View return policy
Specifications
HEK293 | |
ANKRA2 HEK293 Cell Transient Overexpression Lysate(Non-Denatured) | |
Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. ;Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. | |
pCMV-ANKRA2 full length | |
100 μg | |
Human | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Immunoprecipitation, Western Blot | |
NM_023039 | |
Theoretical M.W. 34kDa | |
Transient overexpression cell lysate was tested with anti-ANKRA2 antibody (Mfr. No. H00057763-M01) by Western Blot. | |
In modified RIPA Lysis Buffer. | |
2 mg/mL |