Learn More
Invitrogen™ Annexin A2 Monoclonal Antibody (M298)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA547646
Product Code. 17846908
  • 548.00 € / 100µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
This antibody detects a 36 kDa* protein corresponding to the molecular mass of Annexin A2 on SDS-PAGE immunoblots of rat PC12 cells. The antibody has been shown to label Annexin A2 in immunocytochemical analysis of PC12 cells and immunohistochemical analysis of mouse diaphragm.
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions as an autocrine factor which heightens osteoclast formation and bone resorption. This gene has three pseudogenes located on chromosomes 4, 9 and 10, respectively. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Annexin A2
Monoclonal
0.125 mg/mL
PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
P07355, P07356, Q07936
ANXA2
Clone M009 was generated from a proprietary antigen related to human Annexin A2 in MDA-MB-231 breast cancer cell line.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot, Immunocytochemistry
M298
Unconjugated
ANXA2
33-kDa calcimedin; 33-kDa lymphocyte Ca>2+>- binding protein; 33kDa lymphocyte Ca>2+>- binding protein; 36-kDa calelectrin; annexin 2; annexin A2; annexin A2 isoform 2; annexin II; Annexin-2; ANX2; ANX2L4; ANXA2; AW215814; CAL1H; calpactin I heavy chain; calpactin I heavy chain (p36); calpactin I heavy polypeptide; calpactin-1 heavy chain; chromobindin 8; chromobindin-8; epididymis secretory protein Li 270; HEL-S-270; LIP2; lipocortin II; LPC2; LPC2D; p36; PAP-IV; Placental anticoagulant protein IV; protein I
Mouse
Protein A
RUO
12306, 302, 56611
-20°C, Avoid Freeze/Thaw Cycles