missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTN4, Human, Matched Antibody Pair (AP46), Abnova™
To detect and quantify protein level of human
Brand: Abnova H00000081-AP46.1Set
Additional Details : Weight : 0.00010kg
Includes: Capture antibody: mouse monoclonal anti-ACTN4, IgG1 Kappa (100μg) , Detection antibody: biotinylated mouse monoclonal anti-ACTN4, IgG2b Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
ACTN4 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human ACTN4. | |
ACTININ-4/DKFZp686K23158/FSGS/FSGS1 | |
Mouse | |
RUO | |
81 | |
Human | |
Liquid |
ELISA | |
Biotin | |
ACTN4 | |
ACTN4 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-ACTN4, IgG1 Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-ACTN4, IgG2b Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
For Research Use Only