missing translation for 'onlineSavingsMsg'
Learn More

Cardiac Troponin T Mouse, PE, Clone: 13-11, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 564767

Product Code. 15709144

  • 239.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The 13-11 monoclonal antibody specifically recognizes Troponin T type 2 (cardiac), encoded by the gene TNNT2 . Troponin T is the tropomyosin-binding subunit of the troponin complex, which also encompasses troponin C and troponin I. This complex regulates muscle contraction in skeletal and cardiac muscle in response to alterations in calcium levels. Troponin T type 2 is solely found in the heart, and genetic alterations in the TNNT2 gene are associated to a series of heart disorders in humans, including hypertrophic cardiomyopathy, dilated cardiomyopathy and left ventricular noncompaction. Cardiac Troponin T can be used as a marker for the identification of cardiomyocytes derived from pluripotent stem cells.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cardiac Troponin T
Monoclonal
PE
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Rabbit cardiac troponin T Protein
50 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
13-11
cTnT, Cardiac Muscle Troponin T, Troponin T type 2, TNNT2
Mouse
Affinity Purified
RUO
Canine, Chicken, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.