missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCS, Human, IP-WB Antibody Pair (PW2), Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand: Abnova H00009973-PW2.1Set
Additional Details : Weight : 0.00010kg
Includes: Antibody pair for IP: mouse monoclonal anti-CCS (300μg), Antibody pair for WB: mouse monoclonal anti-CCS (50μg)
Specifications
CCS | |
Monoclonal/Polyclonal | |
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. | |
MGC138260 | |
Mouse | |
RUO | |
9973 | |
Human | |
Liquid |
Immunoprecipitation, Western Blot | |
Unconjugated | |
CCS | |
CCS | |
1 Set | |
Primary | |
Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-CCS (300 ug) 2. Antibody pair for WB: mouse monoclonal anti-CCS (50 ug) | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
For Research Use Only