missing translation for 'onlineSavingsMsg'
Learn More

anti-CDK2, TP53 Protein Protein Interaction Antibody Pair, Abnova™

Brand:  Abnova DI0263.1Set

Product Code. 16191224

  • 648.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: CDK2 rabbit purified polyclonal antibody (20μg); TP53 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
This item is not returnable. View return policy

Description

Description

This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the CDK2 protein, and the other against the TP53 protein for use in in situ Proximity Ligation Assay.

Host Species: Rabbit, Mouse
Species Reactivity: Hu

In situ PLA

Specifications

Specifications

CDK2
Monoclonal/Polyclonal
CDK2
CDK2
1 Set
Primary
Antibody pair set content: 1. CDK2 rabbit purified polyclonal antibody (20 ug) 2. TP53 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Purified
In situ PLA
Unconjugated
p33(CDK2)
Mouse, Rabbit
RUO
1017/7157
Human
CDK2 & TP53 Protein Protein Interaction Antibody Pair
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions