Learn More
Invitrogen™ Coronin 3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530479
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116, NIH-3T3, mouse brain. Predicted reactivity: Mouse (92%), Rat (96%), Chicken (91%), Rhesus Monkey (98%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Coronin 3 is associated with the aggression and metastasis of hepatocellular carcinoma. Coronins are a family of highly evolutionary conserved proteins reportedly involved in the regulation of actin cytoskeletal dynamics, although only coronin 3 has been shown to be related to cancer cell migration.
Specifications
| Coronin 3 | |
| Polyclonal | |
| Unconjugated | |
| Coro1c | |
| AL022675; AW455561; AW548837; Coro1c; coronin 1c; coronin 3; coronin, actin binding protein 1C; coronin, actin binding protein, 1C; coronin, actin-binding protein, 1C; coronin-1C; Coronin-3; CRN2; CRNN4; hCRNN4; RGD1564490 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23603, 23790 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9ULV4, Q9WUM4 | |
| Coro1c | |
| Recombinant protein encompassing a sequence within the C-terminus region of human Coronin 1C. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.