Learn More
anti-MPP1, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand: Abnova H00004354-PW1.1Set
Additional Details : Weight : 0.00010kg
Includes: Antibody pair for IP: mouse monoclonal anti-MPP1 (300μg), Antibody pair for WB: mouse monoclonal anti-MPP1 (50μg)
Specifications
MPP1 | |
Monoclonal/Polyclonal | |
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. | |
AAG12/DXS552/DXS552E/EMP55/MRG1/PEMP | |
Mouse | |
RUO | |
4354 | |
Human | |
Liquid |
Immunoprecipitation, Western Blot | |
Unconjugated | |
MPP1 | |
MPP1 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-MPP1 (300 ug) 2. Antibody pair for WB: mouse monoclonal anti-MPP1 (50 ug) | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
For Research Use Only