Learn More
myosin, light chain 3, alkali; ventricular, skeletal, slow, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human MYL3 protein.
Brand: Abnova H00004634-B01P.50ug
Description
MYL3 encodes myosin light chain 3, an alkali light chain also referred to in the literature as both the ventricular isoform and the slow skeletal muscle isoform. Mutations in MYL3 have been identified as a cause of mid-left ventricular chamber type hypertrophic cardiomyopathy. [provided by RefSeq
Sequence: MAPKKPEPKKDDAKAAPKAAPAPAPPPEPERPKEVEFDASKIKIEFTPEQIEEFKEAFMLFDRTPKCEMKITYGQCGDVLRALGQNPTQAEVLRVLGKPRQEELNTKMMDFETFLPMLQHISKNKDTGTYEDFVEGLRVFDKEGNGTVMGAELRHVLATLGERLTEDEVEKLMAGQEDSNGCINYEAFVKHIMSSSpecifications
| myosin, light chain 3, alkali; ventricular, skeletal, slow | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human MYL3 protein. | |
| MYL3 | |
| CMH8/MLC1SB/MLC1V/VLC1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 4634 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_000258.1 | |
| MYL3 | |
| MYL3 (NP_000249.1, 1 a.a. ∼ 195 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.