Learn More
anti-NMI, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00009111-AP22.1Set
Product Code. 16172964
  • 843.00 € / 1 set
Please to purchase this item. Need a web account? Register with us today!
Includes: Capture antibody: rabbit MaxPab affinity purified polyclonal anti-NMI (100μg) , Detection antibody: mouse purified polyclonal anti-NMI (20μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
NMI
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human NMI.
NMI
1 Set
Primary
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-NMI (100 μg) 2. Detection antibody: mouse purified polyclonal anti-NMI (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
ELISA
Unconjugated
NMI
Mouse, Rabbit
RUO
9111