Learn More
receptor-interacting serine-threonine kinase 4, Mouse, Clone: 1G2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant RIPK4.
Brand: Abnova H00054101-M02.100ug
Description
The protein encoded by this gene is a serine/threonine protein kinase that interacts with protein kinase C-delta. The encoded protein can also activate NFkappaB and is required for keratinocyte differentiation. This kinase undergoes autophosphorylation. [provided by RefSeq
Sequence: HLAARNGHLATVKLLVEEKADVLARGPLNQTALHLAAAHGHSEVVEELVSADVIDLFDEQGLSALHLAAQGRHAQTVETLLRHGAHINLQSLKFQGGHGPAATLLRRSKTSpecifications
| receptor-interacting serine-threonine kinase 4 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_020639 | |
| RIPK4 | |
| RIPK4 (NP_065690, 675 a.a. ∼ 784 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Kinases | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1G2 | |
| Mouse monoclonal antibody raised against a partial recombinant RIPK4. | |
| RIPK4 | |
| ANKK2/ANKRD3/DIK/MGC129992/MGC129993/PKK/RIP4 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 54101 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.