missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-RPS6KA3, PDPK1 Protein Protein Interaction Antibody Pair, Abnova™
Brand: Abnova DI0623.1Set
Includes: RPS6KA3 rabbit purified polyclonal antibody (20μg); PDPK1 mouse purified polyclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Description
Host Species: Rabbit, Mouse
Cross Reactivity: Human
In situ PLA
Specifications
RPS6KA3 | |
Monoclonal/Polyclonal | |
RPS6KA3 | |
RPS6KA3 | |
1 Set | |
6197/5170 | |
Human | |
RPS6KA3 & PDPK1 Protein Protein Interaction Antibody Pair |
In situ PLA | |
Unconjugated | |
CLS/HU-3/ISPK-1/MAPKAPK1B/MRX19/RSK/RSK2/S6K-alpha3/p90-RSK2/pp90RSK2 | |
Mouse, Rabbit | |
Primary | |
Antibody pair set content: 1. RPS6KA3 rabbit purified polyclonal antibody (20 ug) 2. PDPK1 mouse purified polyclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |