Learn More
anti-TNFRSF19, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00055504-AP42.1Set
Product Code. 16143494
  • 809.00 € / 1 set
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Includes: Capture antibody: mouse monoclonal anti-TNFRSF19, IgG1 Kappa (100μg) , Detection antibody: biotinylated mouse monoclonal anti-TNFRSF19, IgG2b Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
TNFRSF19
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TNFRSF19.
TAJ/TAJ-alpha/TRADE/TROY
Mouse
RUO
55504
Human
Liquid
ELISA
Biotin
TNFRSF19
TNFRSF19
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-TNFRSF19, IgG1 Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-TNFRSF19, IgG2b Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.