missing translation for 'onlineSavingsMsg'
Learn More

WTW™ AOX Cell Test Kits

Product Code. 11782265
Click to view available options
Range:
0.05 - 2.50mg/L
Unit Size:
Pack of 25
This item is not returnable. View return policy

Product Code. 11782265

Brand: WTW™ 252023

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

With certificate for optimum precision.

  • Convenient and cost-effective
  • Assured quality by AQA/IQC
  • For infrequent or very large sample throughput in the laboratory
  • Sample adding, inserting, measuring and reading
  • Reaction cuvette with barcode or method selection

TRUSTED_SUSTAINABILITY

Specifications

Analytes Iron(III) thiocyanate
For Use With (Application) For treated sewage, treated electroplating waste water, treated wastewater from chlorine bleaches
For Use With (Equipment) PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral photometers
Includes Certificates
No. of Tests 25
Range 0.05 - 2.50mg/L
Sufficient For 25 Tests
Test Method Photometric
Type AOX cell test kit
compliance-icons compliance-icons compliance-icons
Product Identifier
  • AOX Cell Test Method: photometric 0.05 - 2.50 mg/l Spectroquant
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : methanol, mercury(II) cyanate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.