missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP repeat Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24493-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is used to detect the AP repeat sequence. This antibody detects the APAPAPAPA peptide with dilution 1:32,000 in ELISA.
The C9orf72 "GGGGCC" repeat sequence codes five repeat peptide "GA repeat; GAGAGAGAGA", "GP repeat; GPGPGPGPG", "GR repeat; GRGRGRGRG", "AP repeat; APAPAPAPA" and "PR repeat; PRPRPRPRP". It was described previously that aggregated forms of poly-GA and poly-GP proteins do not enter the separation gel.Specifications
| AP repeat | |
| Polyclonal | |
| Unconjugated | |
| AP repeat, APAPAPAPA, C9RAN, GGGCC, poly AP | |
| Peptide | |
| 20 μL | |
| Primary | |
| -20°C | |
| Liquid |
| Immunohistochemistry, ELISA, Western Blot, Immunofluorescence | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction