Learn More
Applied Biosystems™ MicroAmp™ Snap-On Optical Film Compression Pad
Optical film compression rad attached to metal frame
Brand: Applied Biosystems™ 4333292
Additional Details : Weight : 0.36000kg
Includes: 9 each. Compliant cover bounded to a metal frame and designed for use on the ABI PRISM 7900HT Sequence Detection System when using the available automation accessory with 96-well plates
Description
Designed for use with Applied Biosystems 7900HT Fast Real-Time System when using automation accessory with 96-well plates.
PCR and Real-Time PCR, Real Time PCR (qPCR)
Order Info
Shipping Condition: Room temperature
Specifications
9 each. Compliant cover bounded to a metal frame and designed for use on the ABI PRISM™ 7900HT Sequence Detection System when using the available automation accessory with 96-well plates | |
Optical | |
7900HT System | |
96-Well Plates | |
Room Temperature |
MicroAmp | |
Compression Pad | |
PCR | |
9 |