Learn More
Hirschmann™ AR-Glass Graduated Pipet
Provides volumetric accuracy for your pipetting needs. Hirschmann™ Clear Glass Graduated Pipettes with Amber Stain Graduation with a cotton plug end as standard, short form for tissue culture.
Brand: Hirschmann™ 1000870
This item is not returnable.
View return policy
Description
- Main point ring graduation
- Cotton plug end
- Class AS, Conformity certified, with dated batch
-
Specifications
B | |
Yes | |
0.1 mL | |
0.15 mL | |
450 ±5 mm |
Soda Lime Glass | |
Amber | |
6 Pack | |
25 mL | |
Graduated Pipet |