Learn More
Hirschmann™ AR-Glass Graduated Pipet
Provides volumetric accuracy for your pipetting needs. Hirschmann™ Clear Glass Graduated Pipettes with Amber Stain Graduation with a cotton plug end as standard, short form for tissue culture.
Brand:  Hirschmann™ 1000870
 View more versions of this product
Product Code. 13127899
  • 142.00 € / Pack of 6
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Main point ring graduation
  • Cotton plug end
  • Class AS, Conformity certified, with dated batch
Specifications
Specifications
B
Yes
0.1 mL
0.15 mL
450 ±5 mm
Soda Lime Glass
Amber
6 Pack
25 mL
Graduated Pipet