Learn More
WTW™ Arsenic Reagent Test Kits
With certificate for optimum precision.
Brand: WTW™ 252063
484.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Convenient and cost-effective
- Assured quality by AQA/IQC
- For low concentrations and routine work with large sample throughput in the laboratory
- Wide measuring range using rectangular cuvettes for determination of trace concentrations
Specifications
Phosphorus-molybdenum blue | |
For PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers | |
30 | |
30 Tests | |
Phosphate (PO4) reagent test kit |
For ground water, surface water, drinking water, wastewater | |
Certificates | |
0.001 - 0.100mg/L | |
Photometric |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WTW™ Arsenic Reagent Test Kits > Reagent Test; Detection Range: 0.001 to 0.10mg/L
Spot an opportunity for improvement?Share a Content Correction