missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASB6 293T Cell Overexpression Lysate 1 (Denatured), Abnova
Brand: Abnova H00140459-T01.100uL
This item is not returnable.
View return policy
Description
Western Blotting
Specifications
293T | |
ASB6 293T Cell Transient Overexpression Lysate(Denatured) | |
21.78 | |
pCMV-ASB6 full-length | |
100 μL | |
Human |
Western Blot | |
BC001719 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-ASB6 antibody (H00140459-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 100UL ASB6 293T Cell Transient Overexpression Lysa
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : Mercaptoethanol