Learn More
Thermo Scientific™ Assembled Caps and Septa for 8-425 Standard-Opening Vials
Product Code. 12450148
Click to view available options
Color:
Black
Blue
Natural
Red
White
Yellow
Unit Size:
Pack of 100
Product Code. 12450148
Brand: Thermo Scientific™ C401371A
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ National Assembled Caps and Septa for 8-425 Standard-Opening Vials are available in a variety of colors and septum materials.
Thermo Scientific™ National Assembled Caps and Septa for 8-425 Standard-Opening Vials are available in a variety of colors and septum materials for a wide range of applications.
  • Open top caps can be used with any 8mm septa
  • Phenolic caps are suitable for autoclaving and low temperature applications
  • Polypropylene caps are chemically inert and suitable for most chromatography applications
  • Kim-Lok™ cap/septum assemblies feature a molded centering ring to eliminate septum movement during injection
  • PTFE/red rubber septa are economical for many GC applications
  • PTFE/silicone septa are most popular for HPLC and IC applications
  • Pre-slit septa are easier to pierce with thin-gauge needles
TRUSTED_SUSTAINABILITY
Specifications
Description Phenolic, Black; Ivory PTFE/Red Rubber septum
Color Black
Material PTFE, Rubber, Phenolic
Septum Ivory PTFE/Red Rubber
Type Open Top Cap
Closure Size 8-425
For Use With (Equipment) 8-425 Standard Opening Vials