Learn More
Atropine sulfate monohydrate, 97+%, Thermo Scientific Chemicals
Atropine sulfate monohydrate, CAS # 5908-99-6, is a muscarinic receptor antagonist.
46.33 € - 278.39 €
Chemical Identifiers
CAS | 5908-99-6 |
---|---|
Molecular Formula | H2SO4·H2O |
Molecular Weight (g/mol) | 694.85 |
MDL Number | MFCD00074815 |
InChI Key | PVGPXGKNDGTPTD-IJTOKZDFSA-N |
Synonym | atropine sulfate hydrate, atropine sulfate monohydrate, 1r,3s,5s-8-methyl-8-azabicyclo 3.2.1 octan-3-yl 3-hydroxy-2-phenylpropanoate; methane; sulfuric acid |
PubChem CID | 23624044 |
IUPAC Name | [(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 3-hydroxy-2-phenylpropanoate;sulfuric acid;hydrate |
SMILES | CN1C2CCC1CC(C2)OC(=O)C(CO)C3=CC=CC=C3.O.OS(=O)(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10328270
|
Thermo Scientific Acros
226680100 |
10 g | Glass bottle |
46.33 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10743041
|
Thermo Scientific Acros
226680250 |
25 g | Glass bottle |
89.55 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10464525
|
Thermo Scientific Acros
226681000 |
100 g | Glass bottle |
278.39 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5908-99-6 | |
694.85 | |
PVGPXGKNDGTPTD-IJTOKZDFSA-N | |
23624044 | |
CN1C2CCC1CC(C2)OC(=O)C(CO)C3=CC=CC=C3.O.OS(=O)(=O)O |
H2SO4·H2O | |
MFCD00074815 | |
atropine sulfate hydrate, atropine sulfate monohydrate, 1r,3s,5s-8-methyl-8-azabicyclo 3.2.1 octan-3-yl 3-hydroxy-2-phenylpropanoate; methane; sulfuric acid | |
[(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 3-hydroxy-2-phenylpropanoate;sulfuric acid;hydrate |
Specifications
Passes Test | |
>187.0°C | |
0.2% max. | |
97+% | |
MFCD00074815 | |
15, 866 | |
atropine sulfate hydrate, atropine sulfate monohydrate, 1r,3s,5s-8-methyl-8-azabicyclo 3.2.1 octan-3-yl 3-hydroxy-2-phenylpropanoate; methane; sulfuric acid | |
PVGPXGKNDGTPTD-IJTOKZDFSA-N | |
[(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 3-hydroxy-2-phenylpropanoate;sulfuric acid;hydrate | |
23624044 | |
97+% | |
Atropine sulfate monohydrate |
5908-99-6 | |
White | |
Authentic | |
H2SO4·H2O | |
21, 27 | |
− 0.60 | |
Solubility in water: soluble. Other solubilities: soluble in boiling alcohol and glycerol, slightly soluble in chloroform, practically insoluble in ether | |
CN1C2CCC1CC(C2)OC(=O)C(CO)C3=CC=CC=C3.O.OS(=O)(=O)O | |
694.85 | |
694.85 | |
Powder |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Fatal if swallowed.
Causes skin irritation.
Fatal if inhaled.
May cause an allergic skin reaction.
Causes serious eye irritation.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- Atropine sulfate monohydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
Hazard Statement
- H300-Fatal if swallowed.
- H330-Fatal if inhaled.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
RUO – Research Use Only