missing translation for 'onlineSavingsMsg'
Learn More

AX-024 hydrochloride, MedChemExpress

Product Code. p-200097310
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16492956

Brand: Medchem Express HY107390A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AX-024 hydrochloride is an orally available, first-in-class inhibitor of the TCR-Nck interaction that selectively inhibits TCR-triggered T cell activation with an IC50 ∼1 nM. AX-024 hydrochloride modulates cell signaling by targeting SH3 domains. AX-024 hydrochloride has low-acute toxicity and high potency and selectivity, and strongly inhibit the production of IL-6, TNF-α, IFN-γ, IL-10 and IL-17A.

AX-024 hydrochloride is an orally available, first-in-class inhibitor of the TCR-Nck interaction that selectively inhibits TCR-triggered T cell activation with an IC50 ∼1 nM. AX-024 hydrochloride modulates cell signaling by targeting SH3 domains. AX-024 hydrochloride has low-acute toxicity and high potency and selectivity, and strongly inhibit the production of IL-6, TNF-α, IFN-γ, IL-10 and IL-17A.

Specifications

Chemical Name or Material AX-024 hydrochloride
CAS 1704801-24-0
Color White
Purity Grade Notes Research
Molecular Formula C21H23ClFNO2
Quantity 25 mg
Solubility Information DMSO : 33 mg/mL (87.80 mM; Need ultrasonic and warming) ∣H2O : 16.67 mg/mL (44.35 mM; Need ultrasonic)
SMILES COC1=CC=C(OC2)C(C(C3=CC=C(F)C=C3)=C2CN4CCCC4)=C1.[H]Cl
Molecular Weight (g/mol) 375.86
Formula Weight 375.86
Percent Purity 99.12%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.