Learn More
Axygen™ Tip Refill System, Empty Racks
Easy-to-load refill racks for Axygen Tip Refill System. For use with P2/P10 Gilson-Style and 200μL Universal fit inserts.
Brand: Axygen™ RFL-R-1200
This item is not returnable.
View return policy
Description
Axygen™ Tip Refill System contains durable refill rack that can be easily refilled by hand and is autoclavable. These translucent Racks includes color-coded inserts for tip identification and are certified RNase-/DNase free. They are available in different colors and are packaged in bulk or racks.
- RFL-R is suitable for both the 10 μL and 200 μL refill inserts
- RFL-R-1200 is suitable for 1200 μL refill inserts
- Certified to be RNase-/DNase-free
- Nonpyrogenic
- Translucent rack allows tips to be easily seen
- Color-coded insert allows for easy identification of tip
Specifications
Empty Refill Rack with Lid | |
Non-sterile | |
RFL-R-1200 |
Clear | |
P2/P10 Gilson-Style and 200μL Universal fit inserts | |
10 Racks/Pack; 3 Packs/Case |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Axygen™ Tip Refill System, Empty Racks > 10 Racks/Pk, 5 Pk/Cs
Spot an opportunity for improvement?Share a Content Correction