missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bode Chemie™ Bacillol™ Tissues
Bacillol™ Tissues - Alcohol-based rapid disinfectant wipes in a practical dispenser.
19.50 €
Specifications
Product Type | Disinfectant Wipes |
---|---|
Container Type | Pop-up canister |
Disposable | Single-use |
For Use With (Application) | Disinfection of Alcohol-Resistant Surfaces and Medical Devices |
Product Code | Brand | Container Type | Packaging Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Container Type | Packaging Quantity | Price | Quantity & Availability | |||||
16210991
|
Bode Chemie™
9813114 |
Pop-up canister | 80 Wipes | N/A | ||||||
Description
- Ready-to-use disinfectant wipes
- Quickly and comprehensively effective
- Broad material compatibility with alcohol-resistant surfaces
- Aldehyde, dye and perfume free
- Easy removal from practical dispenser box
- Refillable
- Required spectrum of activity SARS-CoV-2: limited virucidal activity
Specifications
Disinfectant Wipes | |
Pop-up canister | |
Single-use | |
Disinfection of Alcohol-Resistant Surfaces and Medical Devices |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bode Chemie™ Bacillol™ Tissues