Learn More
Gibco™ Bacto™ Proteose Peptone No. 3
Product Code. 16299771
Click to view available options
Quantity:
2 kg
10 kg
50 kg
Unit Size:
10kg
2kg
50kg
For Research Use Only. All usage must comply with product instructions.
Product Code. 16299771
Brand: Gibco™ 212230
Please to purchase this item. Need a web account? Register with us today!
For Research Use Only. All usage must comply with product instructions.
Bacto Proteose Peptone No. 3 is used in preparing microbiological and mammalian cell culture media.
  • Bacto Proteose Peptone No. 3 is a member of the Bacto Proteose Peptones, which are enzymatic digests of protein
  • Studies show that no single peptone is the most suitable nitrogen source for every microbiological application
  • These findings led to the development of Bacto Proteose Peptone, Bacto Proteose Peptone No. 2, and Bacto Proteose Peptone No. 3. Bacto Proteose Peptone No. 4 is a spray-dried version of Bacto Proteose Peptone
  • Bacto Proteose Peptone No. 3 is used in preparing microbiological and mammalian cell culture media
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance LUO, MUO
Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Culture Type Mammalian, Insect, Microbial
Form Powder
Product Type Bacterial Media
Format Pail
For Use With (Application) Enzymatic digest of Protein
Product Line Bacto
Quantity 10 kg
Source Animal Origin