missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ Barbed Elbow/tee/Y-connector Kits, Nylon
Click to view available options
Description:
Barb Connection, Large
Barb Connection, Small
Unit Size:
Each
Description
- Designed for quick and easy assembly without tools.
- Combine these simple and economical fittings with flexible tubing.
- For use in low-pressure applications (generally less than 30psi).
Specifications
Specifications
| Description | Barb Connection, Large |
| Material | Nylon |
| Includes | Elbows: 1/4 in., 3/8 in., 1/2 in., 5/8 in., 3/4 in., 1 in. Tees: 1/4 in., 3/8 in., 1/2 in., 5/8 in., 3/4 in. |
| Product Type | Barbed elbow/tee/Y-connector kits, Nylon, fittings kits (set of 1) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction