missing translation for 'onlineSavingsMsg'
Learn More

Barium Sulfate, 99%, ReagentPlus™, Honeywell™

Product Code. p-7120217 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle
Unit Size:
100g
1kg
500g
This item is not returnable. View return policy

Product Code. 15672600

Brand: Honeywell Research Chemicals 243353100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-43-7
Molecular Formula BaO4S
Molecular Weight (g/mol) 233.38
MDL Number MFCD00003455
InChI Key TZCXTZWJZNENPQ-UHFFFAOYSA-L
Synonym barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
PubChem CID 24414
SMILES [Ba++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1580°C
Color White
Quantity 100 g
Linear Formula BaSO4
UN Number NONH for all modes of transport
IUPAC Name barium(2+) sulfate
Formula Weight 233.39g/mol
Percent Purity 99%
Grade ReagentPlus
Packaging Poly Bottle
Appearance Powder
Density 4.4g/cm3
Chemical Name or Material Barium Sulfate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.