Learn More
Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables
Customize your water purification system with the Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables that best suit your application.
132.00 € - 884.04 €
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10378343
|
Thermo Scientific™
AY1332X1 |
External Booster Pump | N/A | ||||||
10317863
|
Thermo Scientific™
AY1332X2 |
Pre-filtration Accessory Kit |
698.00 €
Each |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||||
11767288
|
Thermo Scientific™
CV742X5A |
Ventgard Reservoir Cap | N/A | ||||||
10156122
|
Thermo Scientific™
D502132 |
Start-up Kit |
776.90 €
1 set |
In Stock
Log in to see stock. |
|||||
10698333
|
Thermo Scientific™
D502133 |
Cartridge Kit |
425.40 €
Each |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
10433893
|
Thermo Scientific™
D7034 |
Empty Cartridges |
208.00 €
Each |
In Stock
Log in to see stock. |
|||||
10235983
|
Thermo Scientific™
LMX13 |
Replacement Ultraviolet Lamp |
257.90 €
Each |
In Stock
Log in to see stock. |
|||||
Description
Customize your water purification system with the Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables that best suit your application. Cartridge kit includes pre-filter, ultrapure mixed bed cartridge and Easypure high purity/low TOC cartridge. Other consumables include 0.2 micron final filter, ultraviolet lamp, Thermo Scientific™ Ventgard™ reservoir cap, reverse osmosis membrane, and set of three empty cartridges for cleaning.
Specifications
Barnstead Easypure RoDi |