Learn More
Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems Replacement UV Lamps
Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
GenPure (UV, UV/UF, UV-TOC and UF/UV-TOC Only), LabTower EDI
LabTower RO
MicroPure, Smart2Pure (UV and UV/UVF Only)
Pacific TII; LabTower TII
Unit Size:
Each
Description
Maintain UV function with Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems Replacement UV Lamps, for use with Barnstead point of use water purification systems.
Ideal for use with the following Barnstead point of use water purification systems:
09.1002:
- MicroPure™ (UV and UV/UVF systems only)
- Smart2Pure™ (UV and UV/UVF systems only)
09.2002:
- GenPure™ (UV, UV/UF, UV-TOC and UF/UV-TOC systems only)
- GenPure™ Pro (UV, UV/UF, UV-TOC and UF/UV-TOC systems only)
- GenPure™ xCAD (UV, UV/UF, UV-TOC and UF/UV-TOC systems only)
- LabTower™ EDI
09.4002:
- Pacific™ TII
- LabTower™ TII
09.5002:
For storage reservoirs (230V only) used with the following Barnstead water purification systems:
- LabTower RO
- LabTower TII
- Pacific TII (30 L, 60 L, 100 L only)
- Smart2Pure (12 only)
Specifications
Specifications
| Description | Replacement UV-Lamp |
| For Use With (Equipment) | LabTower RO |
| Type | UV Lamp |