Learn More
Thermo Scientific™ Barnstead™ RO and TII Accessories and Consumables
Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
Type:
Cartridge, Carbon
Cartridge, MPS
Cartridge, TII Filter
Control Panel Mounting Assembly
Prefilter
Remote Dispenser
Reverse Osmosis Membrane
Reverse Osmosis Mixing Valve
Start-up Kit
Volumetric Remote Dispenser
missing translation for 'unitSize'
Each
Description
Optimize your water purification system with Thermo Scientific™ Barnstead™ RO and TII Accessories and Consumables. For use with Barnstead RO and TII water purification systems, these products include filters and pre-filters, cartridges, overflow cutoff float and pump interlock cable, as well as dispensers and printer.
Optimize your water purification system with Thermo Scientific™ Barnstead™ RO and TII Accessories and Consumables. For use with Barnstead RO and TII water purification systems, these products include filters and pre-filters, cartridges, overflow cutoff float and pump interlock cable, as well as dispensers and printer.
Specifications
Specifications
| For Use With (Equipment) | Barnstead RO |
| Type | Control Panel Mounting Assembly |