missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Dehydrated Culture Media: Orange Serum Agar

Product Code. 11778123
Click to view available options
:
Orange Serum Agar; 500g
This item is not returnable. View return policy

Product Code. 11778123

Brand: BD 211486

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For enumeration and cultivation of microorganisms in citrus juice concentrates and other products

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, COMPF
Content And Storage Refrigerate
Form Dehydrated
Media Type Orange Serum Agar
Product Type Agar
Formulation Orange Serum 10g/L, Yeast extract 3g/L, Pancreatic digest of Casein 10g/L, Dextrose 4g/L, Dipotassium Phosphate 2.5g/L, Agar 15.5g/L
For Use With (Application) Cultivation of aciduric microorganisms, particularly those associated with spoilage of citrus products
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.