missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Mycoflask™ Urea Agar Slants, Complete (Christensen) Prepared Media
Used for the differentiation of organisms, especially the Enterobacteriaceae, on the basis of urease production
55.15 € - 350.00 €
Specifications
Certifications/Compliance | CE, BAM, CCAM, ISO, USDA |
---|---|
Color | Yellow |
Content And Storage | 2°C to 8°C |
Form | Liquid |
Manufacturing Origin | United States |
Description
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
CE, BAM, CCAM, ISO, USDA | |
2°C to 8°C | |
United States | |
Pancreatic digest of Gelatin 1g/L, Dextrose 1g/L, Sodium Chloride 5g/L, Potassium Phosphate 2g/L, Urea 20g/L, Phenol red 0.012g/L, Agar 15g/L | |
Glass Tube with Cap |
Yellow | |
Liquid | |
Urea Agar | |
For differentiation of organisms, especially Enterobacteriaceae, on basis of Urease production |
Safety and Handling
missing translation for 'shelfLife' : 365 Days
For in vitro diagnostic use.