missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Candida Bromcresol Green (BCG) Agar Base
For primary isolation and identification of Candida sp
Brand: BD 283510
This item is not returnable.
View return policy
Description
Specifications
<30°C | |
Candida Bromcresol Green Agar Base | |
Peptone 10g/L, Yeast extract 1g/L, Dextrose 40g/L, Agar 15g/L, Bromcresol green 0.02g/L | |
Bottle |
Dehydrated | |
Agar | |
For primary isolation and identification of Candida sp | |
500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD Difco™ Dehydrated Culture Media: Candida Bromcresol Green (BCG) Agar Base > Candida Bromcresol Green (BCG) Agar Base; 500g
Spot an opportunity for improvement?Share a Content Correction